Assay Details
Target Gene Details
Entrez Gene ID: | 114570 |
Gene Name: | cysteine-rich protein 3 |
Gene Aliases: |
TLP, TLP-A, TLP-B, Tlmp |
Location: |
Chr.17:46428195-46431780 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crip3 | NM_053250.2 | NP_444480.1 | ||
| NM_181664.2 | NP_858050.1 | |||
| AF367970.1 | AAK60524.1 | |||
| AF367971.1 | AAK60525.1 | |||
| AK080272.1 | BAC37863.1 | |||
| AY555745.1 | AAS66891.1 | |||
| BC116360.1 | ||||
| BC116361.1 | 3 | 186 |
More Information
Set Membership: |
|

Genomic Map