Assay Details
Target Gene Details
Entrez Gene ID: | 13819 |
Gene Name: | endothelial PAS domain protein 1 |
Gene Aliases: |
HIF-2alpha, HIF2A, HLF, HRF, MOP2, bHLHe73 |
Location: |
Chr.17:86753864-86833410 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epas1 | NM_010137.3 | NP_034267.3 | ||
| AF045160.1 | AAC12871.1 | |||
| AK136911.1 | ||||
| AK141185.1 | ||||
| AK158430.1 | BAE34506.1 | |||
| BC057870.1 | AAH57870.1 | |||
| D89787.1 | BAA20130.1 | |||
| U81983.1 | AAB41496.1 |
More Information
Set Membership: |
|

Genomic Map