Assay Details
Target Gene Details
Entrez Gene ID: | 268970 |
Gene Name: | Rho GTPase activating protein 28 |
Gene Aliases: |
AU044757, AW550892, E130310N06 |
Location: |
Chr.17:67842706-68004108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap28 | NM_172964.4 | NP_766552.3 | ||
| AK087523.1 | BAC39911.1 | |||
| AK133720.1 | BAE21798.1 | |||
| AK141901.1 | BAE24877.1 | |||
| AK162051.1 | BAE36698.1 | |||
| AK173147.1 | BAD32425.1 | |||
| BC066788.1 | AAH66788.1 |
More Information
Set Membership: |
|

Genomic Map