Assay Details
Target Gene Details
Entrez Gene ID: | 12449 |
Gene Name: | cyclin F |
Gene Aliases: |
CycF, Fbxo1 |
Location: |
Chr.17:24223230-24251409 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnf | NM_007634.4 | 10 | 1069 | NP_031660.3 |
| AK032961.1 | 10 | 995 | ||
| AK082485.1 | 10 | 1037 | BAC38507.1 | |
| AK085183.1 | 10 | 1006 | ||
| AK159594.1 | 10 | 1070 | BAE35214.1 | |
| AK162447.1 | 10 | 1009 | ||
| AK169263.1 | 10 | 1020 | BAE41025.1 | |
| AK215309.1 | 3 | 238 | ||
| BC037662.1 | 10 | 1015 | AAH37662.1 | |
| U20612.1 | 10 | 1007 | AAA62317.1 | |
| U20636.1 | 10 | 1124 | AAA63152.1 | |
| Z47766.1 | 10 | 1009 | CAA87695.1 |
Target Gene Details
Entrez Gene ID: | 100628603 |
Gene Name: | microRNA 5134 |
Gene Aliases: |
mir-5134, mmu-mir-5134, mu-mir-5134 |
Location: |
Chr.17:24234527-24234604 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir5134 | NR_039596.1 | |||
| JF446437.1 |
More Information
Set Membership: |
|

Genomic Map