Assay Details
Target Gene Details
Entrez Gene ID: | 11571 |
Gene Name: | cysteine-rich secretory protein 1 |
Gene Aliases: |
AW324342, Aeg1, CRISP-1 |
Location: |
Chr.17:40293758-40319246 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crisp1 | NM_009638.3 | NP_033768.3 | ||
| AK009449.1 | BAB26295.1 | |||
| BC011150.1 | AAH11150.1 | |||
| L05559.1 | AAA37460.1 | |||
| M92849.1 | AAA37185.1 |

Genomic Map