Assay Details
Target Gene Details
Entrez Gene ID: | 22024 |
Gene Name: | cysteine-rich secretory protein 2 |
Gene Aliases: |
CRISP-2, GAPDL5, Tpx-1, Tpx1 |
Location: |
Chr.17:40764734-40794146 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crisp2 | NM_001204071.1 | 7 | 592 | NP_001191000.1 |
| NM_009420.2 | 7 | 610 | NP_033446.1 | |
| BC049615.1 | 7 | 562 | AAH49615.1 | |
| M25533.1 | 7 | 562 | AAA40472.1 |
More Information
Set Membership: |
|

Genomic Map