Assay Details
Target Gene Details
Entrez Gene ID: | 13388 |
Gene Name: | delta-like 1 (Drosophila) |
Gene Aliases: |
Delta1 |
Location: |
Chr.17:15367354-15376113 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dll1 | NM_007865.3 | NP_031891.2 | ||
| BC057400.1 | AAH57400.1 | |||
| BC065063.1 | AAH65063.1 | |||
| X80903.1 | CAA56865.1 |

Genomic Map