Assay Details
Target Gene Details
Entrez Gene ID: | 13388 |
Gene Name: | delta-like 1 (Drosophila) |
Gene Aliases: |
Delta1 |
Location: |
Chr.17:15367354-15376113 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dll1 | NM_007865.3 | 8 | 1687 | NP_031891.2 |
| BC057400.1 | 8 | 1200 | AAH57400.1 | |
| BC065063.1 | 8 | 1451 | AAH65063.1 | |
| X80903.1 | 8 | 1117 | CAA56865.1 |

Genomic Map