Assay Details
Target Gene Details
Entrez Gene ID: | 57440 |
Gene Name: | EH-domain containing 3 |
Gene Aliases: |
Ehd2 |
Location: |
Chr.17:73804841-73832093 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ehd3 | NM_020578.3 | NP_065603.2 | ||
| AF155883.1 | AAF19020.1 | |||
| AF506002.1 | AAM28633.1 | |||
| AK128964.1 | ||||
| AK141265.1 | BAE24623.1 | |||
| BC046596.1 | AAH46596.1 |
More Information
Set Membership: |
|

Genomic Map