Assay Details
Target Gene Details
Entrez Gene ID: | 68655 |
Gene Name: | fibronectin type III domain containing 1 |
Gene Aliases: |
1110027O12Rik, 1110051A18Rik, mKIAA1866 |
Location: |
Chr.17:7738568-7827342 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fndc1 | NM_001081416.1 | NP_001074885.1 |

Genomic Map