Assay Details
Target Gene Details
Entrez Gene ID: | 66968 |
Gene Name: | perilipin 5 |
Gene Aliases: |
2310076L09Rik, AI415325, AW109675, Lsdp5, MLDP, PAT-1 |
Location: |
Chr.17:56111597-56117606 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plin5 | NM_001077348.1 | NP_001070816.1 | ||
| NM_025874.3 | NP_080150.2 | |||
| AK010202.1 | ||||
| AK075872.1 | BAC36019.1 | |||
| AY919875.1 | AAX12443.1 | |||
| BC024138.1 | ||||
| DQ473305.1 |
More Information
Set Membership: |
|

Genomic Map