Assay Details
Target Gene Details
Entrez Gene ID: | 78785 |
Gene Name: | CAP-GLY domain containing linker protein family, member 4 |
Gene Aliases: |
1700024K14Rik, 1700074B05Rik, 4833417L20Rik, 5830409B12Rik, Rsnl2 |
Location: |
Chr.17:71769669-71864210 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clip4 | NM_001271483.1 | 4 | 460 | NP_001258412.1 |
| NM_001271484.1 | 4 | 545 | NP_001258413.1 | |
| NM_030179.3 | 4 | 448 | NP_084455.2 | |
| NM_175378.1 | 4 | 545 | NP_780587.1 | |
| NR_073185.1 | 4 | 545 | ||
| AK017914.1 | 1 | 35 | ||
| AK018895.1 | 4 | 538 | BAB31478.1 | |
| AK019516.1 | 4 | 461 | ||
| AK029962.1 | 4 | 546 | ||
| AK075758.1 | 4 | 536 | BAC35936.1 | |
| AK161301.1 | 4 | 535 | BAE36308.1 | |
| BC035226.1 | 4 | 448 | AAH35226.1 |
More Information
Set Membership: |
|

Genomic Map