Assay Details
Target Gene Details
Entrez Gene ID: | 13163 |
Gene Name: | Fas death domain-associated protein |
Gene Aliases: |
- |
Location: |
Chr.17:33909381-33915590 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daxx | NM_001199733.1 | NP_001186662.1 | ||
| NM_007829.4 | NP_031855.3 | |||
| AF006040.1 | AAC53284.1 | |||
| AK090356.1 | ||||
| AK133863.1 | BAE21895.1 | |||
| AK144332.1 | ||||
| AK145906.1 | BAE26741.1 | |||
| AK146743.1 | BAE27403.1 | |||
| AK151279.1 | ||||
| AK153149.1 | BAE31759.1 | |||
| AK156425.1 | BAE33708.1 | |||
| AK172616.1 | ||||
| AY734459.1 | AAU87500.1 | |||
| AY734462.1 | AAU50681.1 | |||
| AY734465.1 | AAU45095.1 | |||
| AY758273.1 | AAU84992.1 | |||
| BC128373.1 | ||||
| KJ650207.1 |

Genomic Map