Assay Details
Target Gene Details
Entrez Gene ID: | 13163 |
Gene Name: | Fas death domain-associated protein |
Gene Aliases: |
- |
Location: |
Chr.17:33909381-33915590 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daxx | NM_001199733.1 | 8 | 2514 | NP_001186662.1 |
| NM_007829.4 | 8 | 2414 | NP_031855.3 | |
| AF006040.1 | 7 | 2265 | AAC53284.1 | |
| AK090356.1 | 9 | 2225 | ||
| AK133863.1 | 8 | 2322 | BAE21895.1 | |
| AK144332.1 | 8 | 2347 | ||
| AK145906.1 | 8 | 2345 | BAE26741.1 | |
| AK146743.1 | 8 | 2341 | BAE27403.1 | |
| AK151279.1 | 8 | 2335 | ||
| AK153149.1 | 8 | 2302 | BAE31759.1 | |
| AK156425.1 | 8 | 2512 | BAE33708.1 | |
| AK172616.1 | 8 | 2515 | ||
| AK209646.1 | 1 | 56 | ||
| AY734459.1 | AAU87500.1 | |||
| AY734464.1 | AAU50683.1 | |||
| AY734465.1 | 6 | 1889 | AAU45095.1 | |
| AY758274.1 | 3 | 720 | AAU84993.1 | |
| BC128373.1 | 8 | 2324 | ||
| BQ176437.1 | 1 | 185 | ||
| KJ650207.1 |

Genomic Map