Assay Details
Target Gene Details
Entrez Gene ID: | 214505 |
Gene Name: | N-acetylglucosamine-1-phosphotransferase, gamma subunit |
Gene Aliases: |
6430527N14Rik, A830081F19, AU067667, AU067744, Mdcp1, Tce7 |
Location: |
Chr.17:25234317-25240170 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnptg | NM_172529.3 | NP_766117.2 | ||
| AK044021.1 | ||||
| AY465529.1 | AAR19081.1 | |||
| BC055872.1 | AAH55872.1 |
More Information
Set Membership: |
|

Genomic Map