Assay Details
Target Gene Details
Entrez Gene ID: | 15024 |
Gene Name: | histocompatibility 2, T region locus 10 |
Gene Aliases: |
H-2T10 |
Location: |
Chr.17:36115871-36121444 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-T10 | NR_046286.1 | 4 | 698 | |
| AK088241.1 | ||||
| AK161731.1 | 2 | 988 | ||
| M35244.1 | 3 | 787 | AAA39695.1 | |
| M35246.1 | 4 | 664 | AAA39694.1 |
More Information
Set Membership: |
|

Genomic Map