Assay Details
Target Gene Details
Entrez Gene ID: | 16580 |
Gene Name: | kinesin family member C5B |
Gene Aliases: |
Hset, KNSL2, Kifc5a |
Location: |
Chr.17:26917091-26932579 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kifc5b | NM_053173.2 | 1 | 52 | NP_444403.2 |
| AK041901.1 | 1 | 53 | ||
| BB638629.1 | 1 | 53 | ||
| BC091570.1 | AAH91570.1 |
More Information
Set Membership: |
|

Genomic Map