Assay Details
Target Gene Details
Entrez Gene ID: | 106628 |
Gene Name: | thyroid hormone receptor interactor 10 |
Gene Aliases: |
AI646975, Cip4 |
Location: |
Chr.17:57233207-57263697 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip10 |

Genomic Map