Assay Details
Target Gene Details
Entrez Gene ID: | 225028 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 3 |
Gene Aliases: |
4833416M01Rik, 4833416M07Rik, 9530052P13Rik, Glk, MAPKKKK3, MEKKK 3, MEKKK3, RAB8IPL1 |
Location: |
Chr.17:80580513-80728205 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map4k3 | NM_001290345.1 | NP_001277274.1 | ||
| BC005781.1 | AAH05781.1 | |||
| CD353921.1 | ||||
| CO433380.1 |

Genomic Map