Assay Details
Target Gene Details
Entrez Gene ID: | 18763 |
Gene Name: | polycystic kidney disease 1 homolog |
Gene Aliases: |
PC1, mFLJ00285 |
Location: |
Chr.17:24549950-24596514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkd1 | NM_013630.2 | NP_038658.2 | ||
| U70209.1 | AAC53207.1 |
More Information
Set Membership: |
|

Genomic Map