Assay Details
Target Gene Details
Entrez Gene ID: | 22092 |
Gene Name: | radial spoke head 1 homolog (Chlamydomonas) |
Gene Aliases: |
MCA, Tsga2 |
Location: |
Chr.17:31255019-31277356 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsph1 | NM_025290.3 | 7 | 756 | NP_079566.1 |
| AB006535.1 | 7 | 744 | BAB83693.1 | |
| AK005739.1 | 7 | 708 | BAB24214.1 | |
| BC049584.1 | 7 | 756 | AAH49584.1 | |
| DQ141549.1 | 7 | 655 | AAZ75687.1 | |
| DQ141550.1 | 7 | 655 | AAZ75688.1 | |
| DQ141551.1 | 7 | 655 | AAZ75689.1 | |
| DQ141553.1 | 7 | 655 | AAZ75691.1 | |
| DQ141554.1 | 7 | 655 | AAZ75692.1 | |
| DQ141555.1 | 6 | 541 | AAZ75693.1 | |
| DQ141556.1 | 6 | 541 | AAZ75694.1 |

Genomic Map