Assay Details
Target Gene Details
Entrez Gene ID: | 19326 |
Gene Name: | RAB11B, member RAS oncogene family |
Gene Aliases: |
A730055L17Rik |
Location: |
Chr.17:33742484-33760486 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11b | NM_008997.3 | NP_033023.1 | ||
| AB232606.1 | BAF02868.1 | |||
| AK134125.1 | BAE22024.1 | |||
| AK146452.1 | BAE27183.1 | |||
| AK146460.1 | BAE27188.1 | |||
| AK146623.1 | BAE27312.1 | |||
| AK146978.1 | BAE27582.1 | |||
| AK147077.1 | BAE27658.1 | |||
| AK148022.1 | BAE28293.1 | |||
| AK148106.1 | BAE28347.1 | |||
| AK188721.1 | ||||
| AK203890.1 | ||||
| AK205000.1 | ||||
| BC054753.1 | AAH54753.1 | |||
| BC085270.1 | AAH85270.1 | |||
| BC157916.1 | ||||
| BU705914.1 | ||||
| KJ650201.1 | ||||
| L26528.1 | AAC42093.1 |

Genomic Map