Assay Details
Target Gene Details
Entrez Gene ID: | 26415 |
Gene Name: | mitogen-activated protein kinase 13 |
Gene Aliases: |
SAPK4, Serk4 |
Location: |
Chr.17:28769317-28778704 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk13 | NM_011950.2 | 11 | 978 | NP_036080.2 |
| AK146993.1 | 11 | 966 | BAE27594.1 | |
| BC001992.1 | 11 | 947 | AAH01992.1 | |
| U81823.1 | 11 | 935 | AAD09229.1 |
More Information
Set Membership: |
|

Genomic Map