Assay Details
Target Gene Details
Entrez Gene ID: | 17929 |
Gene Name: | myomesin 1 |
Gene Aliases: |
D430047A17Rik |
Location: |
Chr.17:71019002-71126856 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myom1 | NM_001083934.1 | 6 | 1038 | NP_001077403.1 |
| NM_010867.2 | 6 | 1038 | NP_034997.2 | |
| AJ012072.1 | 6 | 1071 | CAB46494.1 | |
| AK052539.1 | 6 | 1005 | ||
| BC060378.1 | 5 | 943 | AAH60378.1 | |
| Z22866.1 | 6 | 1017 | CAA80488.1 |

Genomic Map