Assay Details
Target Gene Details
Entrez Gene ID: | 67242 |
Gene Name: | gem (nuclear organelle) associated protein 6 |
Gene Aliases: |
2610019B15Rik, 2810470M17Rik |
Location: |
Chr.17:80207704-80228497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gemin6 | NM_026053.3 | NP_080329.1 | ||
| AK011454.1 | BAB27627.1 | |||
| AK013396.1 | BAB28830.1 | |||
| AK018385.1 | BAB31187.1 | |||
| AK020147.1 | BAB32011.1 | |||
| AK157745.1 | BAE34178.1 | |||
| AK167680.1 | BAE39729.1 | |||
| AK167920.1 | BAE39927.1 | |||
| BC025157.1 | AAH25157.1 |

Genomic Map