Assay Details
Target Gene Details
Entrez Gene ID: | 11692 |
Gene Name: | growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration) |
Gene Aliases: |
Alr, ERV1 |
Location: |
Chr.17:24693189-24696227 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfer | NM_023040.3 | 2 | 389 | NP_075527.2 |
| AB041561.1 | 2 | 306 | BAA95045.1 | |
| AF148688.1 | 2 | 169 | AAD36987.1 | |
| AK146579.1 | 2 | 390 | BAE27275.1 | |
| BC023941.1 | 2 | 389 | AAH23941.1 | |
| BI901357.1 | 2 | 400 |
More Information
Set Membership: |
|

Genomic Map