Assay Details
Target Gene Details
Entrez Gene ID: | 68977 |
Gene Name: | hydroxyacylglutathione hydrolase-like |
Gene Aliases: |
1500017E18Rik, 2810014I23Rik, C330022E15Rik |
Location: |
Chr.17:25779843-25785669 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Haghl | NM_001271433.1 | NP_001258362.1 | ||
| NM_001271434.1 | NP_001258363.1 | |||
| NM_001271435.1 | NP_001258364.1 | |||
| NM_001271436.1 | NP_001258365.1 | |||
| NM_001271437.1 | NP_001258366.1 | |||
| NM_026897.3 | NP_081173.1 | |||
| AK005274.1 | BAB23924.1 | |||
| AK012748.1 | ||||
| AK021220.1 | ||||
| AK032999.1 | ||||
| AK034798.1 | ||||
| AK047517.1 | BAC33079.1 | |||
| AK079632.1 | ||||
| AK080454.1 | 1 | 1985 | ||
| AK154248.1 | ||||
| AK185790.1 | ||||
| BC030466.1 | AAH30466.1 | |||
| BC083322.1 | AAH83322.1 |

Genomic Map