Assay Details
Target Gene Details
Entrez Gene ID: | 353502 |
Gene Name: | host cell factor C1 regulator 1 (XPO1-dependent) |
Gene Aliases: |
HPIP |
Location: |
Chr.17:23673608-23675524 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcfc1r1 | NM_181821.1 | NP_861542.1 | ||
| AK003865.1 | ||||
| AK013438.1 | BAB28855.1 | |||
| BC079865.1 | AAH79865.1 |

Genomic Map