Assay Details
Target Gene Details
Entrez Gene ID: | 16772 |
Gene Name: | laminin, alpha 1 |
Gene Aliases: |
AA408497, Lama |
Location: |
Chr.17:67697133-67822645 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lama1 | NM_008480.2 | 1 | 32 | NP_032506.2 |
| AK051116.1 | 1 | 33 | ||
| J04064.1 | AAA39410.1 | |||
| X07737.1 | 1 | 39 | CAA30561.1 |
More Information
Set Membership: |
|

Genomic Map