Assay Details
Target Gene Details
Entrez Gene ID: | 109801 |
Gene Name: | glyoxalase 1 |
Gene Aliases: |
0610009E22Rik, 1110008E19Rik, 2510049H23Rik, AW550643, GLY1, Glo-1, Glo-1r, Glo-1s, Glo1-r, Glo1-s, Qglo |
Location: |
Chr.17:30592861-30612659 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glo1 | NM_001113560.1 | 7 | 790 | NP_001107032.1 |
| NM_025374.3 | 6 | 1830 | NP_079650.3 | |
| AK002386.1 | 6 | 1758 | BAB22060.1 | |
| AK003567.1 | 6 | 1778 | BAB22863.1 | |
| AK049703.1 | 6 | 1787 | BAC33882.1 | |
| AK196705.1 | 1 | 172 | ||
| AK202257.1 | 1 | 81 | ||
| BC024663.1 | 6 | 1789 | AAH24663.1 | |
| BP769280.1 | 1 | 129 |
More Information
Set Membership: |
|

Genomic Map