Assay Details
Target Gene Details
Entrez Gene ID: | 109801 |
Gene Name: | glyoxalase 1 |
Gene Aliases: |
0610009E22Rik, 1110008E19Rik, 2510049H23Rik, AW550643, GLY1, Glo-1, Glo-1r, Glo-1s, Glo1-r, Glo1-s, Qglo |
Location: |
Chr.17:30592861-30612659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glo1 | NM_001113560.1 | NP_001107032.1 | ||
| NM_025374.3 | NP_079650.3 | |||
| AK002386.1 | BAB22060.1 | |||
| AK003567.1 | BAB22863.1 | |||
| AK005055.1 | BAB23781.1 | |||
| AK005215.1 | ||||
| AK031832.1 | BAC27570.1 | |||
| AK049703.1 | BAC33882.1 | |||
| BC024663.1 | AAH24663.1 | |||
| BC081432.1 | AAH81432.1 | |||
| BG077218.2 | ||||
| BY312309.1 | ||||
| DQ487161.1 | ||||
| DQ487162.1 |
More Information
Set Membership: |
|

Genomic Map