Assay Details
Target Gene Details
Entrez Gene ID: | 27756 |
Gene Name: | LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae) |
Gene Aliases: |
D17H6S56E-2, D17H6S56E2, Dmapl, Dmpkap, G7b, Sm-X5, SmX5, snRNP |
Location: |
Chr.17:34981854-34985891 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsm2 | NM_001110101.2 | 1 | 169 | NP_001103571.1 |
| NM_001204274.1 | 1 | 169 | NP_001191203.1 | |
| AK151605.1 | 1 | 143 | BAE30544.1 | |
| AK162880.1 | 1 | 161 | ||
| BC014288.1 | 1 | 154 | AAH14288.1 | |
| BY079930.1 | 1 | 171 |
More Information
Set Membership: |
|

Genomic Map