Assay Details
Target Gene Details
Entrez Gene ID: | 27756 |
Gene Name: | LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae) |
Gene Aliases: |
D17H6S56E-2, D17H6S56E2, Dmapl, Dmpkap, G7b, Sm-X5, SmX5, snRNP |
Location: |
Chr.17:34981854-34985891 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsm2 | NM_001110101.2 | NP_001103571.1 | ||
| NM_001204273.1 | 1 | 113 | NP_001191202.1 | |
| NM_001204274.1 | NP_001191203.1 | |||
| NM_030597.3 | 1 | 113 | NP_085100.1 | |
| AF204146.1 | 1 | 84 | AAG31423.1 | |
| AF204147.1 | 1 | 84 | AAG31424.1 | |
| AF204148.1 | 1 | 84 | AAG31425.1 | |
| AF204149.1 | 1 | 84 | AAG31426.1 | |
| AF204150.1 | 1 | 84 | AAG31427.1 | |
| AF204151.1 | 1 | 84 | AAG31428.1 | |
| AF204152.1 | 1 | 84 | AAG31429.1 | |
| AF204153.1 | 1 | 74 | AAG31430.1 | |
| AF204154.1 | 1 | 84 | AAG31431.1 | |
| AF204155.1 | 1 | 84 | AAG31432.1 | |
| AF204156.1 | AAG31433.1 | |||
| AF204157.1 | AAG31434.1 | |||
| AK151605.1 | BAE30544.1 | |||
| AK162880.1 | ||||
| AK178855.1 | ||||
| BB848823.1 | 1 | 113 | ||
| BC014288.1 | AAH14288.1 | |||
| BY079930.1 |
More Information
Set Membership: |
|

Genomic Map