Assay Details
Target Gene Details
Entrez Gene ID: | 17685 |
Gene Name: | mutS homolog 2 (E. coli) |
Gene Aliases: |
AI788990 |
Location: |
Chr.17:87672557-87723713 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msh2 | NM_008628.2 | NP_032654.1 | ||
| AK051960.1 | ||||
| AK157842.1 | BAE34223.1 | |||
| AK166179.1 | ||||
| AK217675.1 | ||||
| BC047117.1 | AAH47117.1 | |||
| BC050897.1 | AAH50897.1 | |||
| U21011.1 | AAA75027.1 | |||
| X81143.1 | CAA57049.1 |
More Information
Set Membership: |
|

Genomic Map