Assay Details
Target Gene Details
Entrez Gene ID: | 268934 |
Gene Name: | glutamate receptor, metabotropic 4 |
Gene Aliases: |
Gprc1d, mGluR4 |
Location: |
Chr.17:27422387-27513679 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm4 | NM_001013385.2 | 3 | 1178 | NP_001013403.1 |
| NM_001291045.1 | 3 | 1178 | NP_001277974.1 | |
| BC080284.1 | 3 | 949 | AAH80284.1 |
More Information
Set Membership: |
|

Genomic Map