Assay Details
Target Gene Details
Entrez Gene ID: | 16440 |
Gene Name: | inositol 1,4,5-triphosphate receptor 3 |
Gene Aliases: |
Ip3r3, Itpr-3 |
Location: |
Chr.17:27057260-27122223 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpr3 | NM_080553.3 | NP_542120.2 | ||
| AB182289.1 | BAD90683.1 |
More Information
Set Membership: |
|

Genomic Map