Assay Details
Target Gene Details
Entrez Gene ID: | 18038 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor like 1 |
Gene Aliases: |
Def-7, IKBL |
Location: |
Chr.17:35220175-35236452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkbil1 | NM_010909.4 | NP_035039.1 | ||
| AF095902.1 | AAC64292.1 | |||
| AK133517.1 | BAE21699.1 | |||
| BC013269.1 | AAH13269.1 |
More Information
Set Membership: |
|

Genomic Map