Assay Details
Target Gene Details
Entrez Gene ID: | 18044 |
Gene Name: | nuclear transcription factor-Y alpha |
Gene Aliases: |
AA407810, Cbf-b, SEZ-10, SEZ10 |
Location: |
Chr.17:48386885-48409901 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfya | NM_001110832.1 | NP_001104302.1 | ||
| NM_010913.2 | NP_035043.1 | |||
| AK004729.1 | BAB23511.1 | |||
| AK137561.1 | 2 | 279 | ||
| AK143723.1 | BAE25515.1 | |||
| AK156487.1 | BAE33729.1 | |||
| BC029695.1 | AAH29695.1 | |||
| BC057099.1 | AAH57099.1 | |||
| BY127163.1 | ||||
| M86214.1 | AAA39816.1 | |||
| M86215.1 | AAA39817.1 | |||
| X55315.1 | CAA39023.1 |
More Information
Set Membership: |
|

Genomic Map