Assay Details
Target Gene Details
Entrez Gene ID: | 19133 |
Gene Name: | peripherin 2 |
Gene Aliases: |
AOFMD, AVMD, Nmf193, PRPH, RP7, Rd-2, Rd2, Rds, Tspan22, rds |
Location: |
Chr.17:46910485-46924926 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prph2 | NM_008938.1 | 2 | 955 | NP_032964.1 |
| AK044549.1 | 2 | 950 | BAC31975.1 | |
| AK136280.1 | 2 | 982 | BAE22911.1 | |
| BC028958.1 | 2 | 950 | AAH28958.1 | |
| BC085266.1 | 2 | 954 | AAH85266.1 | |
| X14770.1 | 2 | 955 | CAA32878.1 |

Genomic Map