Assay Details
Target Gene Details
Entrez Gene ID: | 19765 |
Gene Name: | ralA binding protein 1 |
Gene Aliases: |
RLIP76, Rip1 |
Location: |
Chr.17:65848415-65885755 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralbp1 | NM_001198949.1 | 5 | 1479 | NP_001185878.1 |
| NM_009067.5 | 5 | 1803 | NP_033093.2 | |
| AK136252.1 | 3 | 869 | ||
| AK145387.1 | 5 | 1343 | BAE26405.1 | |
| AK159404.1 | 5 | 1356 | BAE35056.1 | |
| BC067073.1 | 4 | 1238 | AAH67073.1 | |
| BC075732.1 | 5 | 1428 | AAH75732.1 | |
| BC076636.1 | 5 | 1395 | AAH76636.1 | |
| BC138546.1 | 4 | 1244 | ||
| BC145320.1 | ||||
| BC145815.1 | 4 | 1244 | ||
| X80937.1 | 5 | 1380 | CAA56912.1 |
More Information
Set Membership: |
|

Genomic Map