Assay Details
Target Gene Details
Entrez Gene ID: | 80880 |
Gene Name: | KN motif and ankyrin repeat domains 3 |
Gene Aliases: |
0610013D04Rik, Ankrd47, D17Ertd288e, NG28 |
Location: |
Chr.17:33810523-33822920 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kank3 | NM_030697.2 | NP_109622.1 | ||
| AK002623.1 | BAB22238.1 | |||
| AK132011.1 | BAE20938.1 | |||
| BC061055.1 | AAH61055.1 | |||
| BC115933.1 | ||||
| KJ650203.1 | ||||
| KJ663713.1 | ||||
| KJ663714.1 |
More Information
Set Membership: |
|

Genomic Map