Assay Details
Target Gene Details
Entrez Gene ID: | 56055 |
Gene Name: | GTP binding protein 2 |
Gene Aliases: |
nmf205 |
Location: |
Chr.17:46159583-46169370 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp2 | NM_001145979.1 | NP_001139451.1 | ||
| NM_019581.3 | NP_062527.2 | |||
| AB024573.1 | BAB12430.1 | |||
| AF168991.1 | AAF78885.1 | |||
| AK087511.1 | ||||
| AK146415.1 | BAE27151.1 | |||
| AK148020.1 | ||||
| BC049089.1 | AAH49089.1 | |||
| BC051958.1 | AAH51958.1 |
More Information
Set Membership: |
|

Genomic Map