Assay Details
Target Gene Details
Entrez Gene ID: | 56055 |
Gene Name: | GTP binding protein 2 |
Gene Aliases: |
nmf205 |
Location: |
Chr.17:46159583-46169370 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp2 | NM_001145979.1 | 8 | 1247 | NP_001139451.1 |
| NM_019581.3 | 8 | 1282 | NP_062527.2 | |
| AB024573.1 | 6 | 918 | BAB12430.1 | |
| AF168991.1 | 8 | 1187 | AAF78885.1 | |
| AK087511.1 | 8 | 1153 | ||
| AK146415.1 | 8 | 1232 | BAE27151.1 | |
| AK148020.1 | 8 | 1203 | ||
| BC049089.1 | 8 | 1243 | AAH49089.1 | |
| BC051958.1 | 8 | 1243 | AAH51958.1 |
More Information
Set Membership: |
|

Genomic Map