Assay Details
Target Gene Details
Entrez Gene ID: | 56047 |
Gene Name: | mesothelin |
Gene Aliases: |
MPF |
Location: |
Chr.17:25748613-25754437 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msln | NM_018857.1 | 16 | 2048 | NP_061345.1 |
| AK144391.1 | 17 | 1980 | BAE25864.1 | |
| AK146799.1 | 16 | 2098 | BAE27441.1 | |
| BC023753.1 | 16 | 2052 | AAH23753.1 | |
| D86370.1 | 16 | 2048 | BAA13077.1 |
More Information
Set Membership: |
|

Genomic Map