Assay Details
Target Gene Details
Entrez Gene ID: | 240168 |
Gene Name: | RAS, guanyl releasing protein 3 |
Gene Aliases: |
BC066069, Gm327 |
Location: |
Chr.17:75435871-75529054 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasgrp3 | NM_001166493.1 | NP_001159965.1 | ||
| NM_207246.4 | 2 | 179 | NP_997129.1 | |
| AK173041.1 | BAD32319.1 | |||
| BC066127.1 | 2 | 104 | AAH66127.1 | |
| BY123705.1 | 2 | 99 | ||
| CJ064681.1 | ||||
| CJ131259.1 | 2 | 181 | ||
| CN695386.1 |
More Information
Set Membership: |
|

Genomic Map