Assay Details
Target Gene Details
Entrez Gene ID: | 76936 |
Gene Name: | heterogeneous nuclear ribonucleoprotein M |
Gene Aliases: |
2610023M21Rik, AA409009, Hnrpm, mKIAA4193 |
Location: |
Chr.17:33646233-33685458 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpm | NM_001109913.1 | NP_001103383.1 | ||
| NM_029804.3 | NP_084080.1 | |||
| AK011521.1 | BAB27675.1 | |||
| AK048220.1 | 1 | 2134 | ||
| AK133911.1 | ||||
| AK145343.1 | BAE26377.1 | |||
| AK159848.1 | BAE35426.1 | |||
| AK166670.1 | BAE38931.1 | |||
| AK168347.1 | BAE40284.1 | |||
| AK169101.1 | ||||
| AK171892.1 | BAE42723.1 | |||
| AK220299.1 | BAD90224.1 | |||
| BC065172.1 | AAH65172.1 |
More Information
Set Membership: |
|

Genomic Map