Assay Details
Target Gene Details
Entrez Gene ID: | 224824 |
Gene Name: | peroxisomal biogenesis factor 6 |
Gene Aliases: |
AI132582, D130055I09Rik, mKIAA4177 |
Location: |
Chr.17:46711463-46725541 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex6 | NM_145488.1 | NP_663463.1 | ||
| AK143173.1 | ||||
| AK182393.1 | ||||
| AK199420.1 | ||||
| AK220525.1 | BAD90524.1 | |||
| BC003424.1 | AAH03424.1 |

Genomic Map