Assay Details
Target Gene Details
Entrez Gene ID: | 80905 |
Gene Name: | polymerase (DNA directed), eta (RAD 30 related) |
Gene Aliases: |
RAD30A, XPV |
Location: |
Chr.17:46171355-46202683 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polh | NM_030715.3 | NP_109640.1 | ||
| AB027128.1 | BAA97570.1 | |||
| AK036296.1 | BAC29375.1 |
More Information
Set Membership: |
|

Genomic Map