Assay Details
Target Gene Details
Entrez Gene ID: | 18132 |
Gene Name: | notch 4 |
Gene Aliases: |
Int-3, Int3, N4 |
Location: |
Chr.17:34564282-34588543 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Notch4 | NM_010929.2 | NP_035059.2 | ||
| BC138044.1 | ||||
| M80456.1 | AAB38377.1 |
More Information
Set Membership: |
|

Genomic Map