Assay Details
Target Gene Details
Entrez Gene ID: | 72836 |
Gene Name: | protection of telomeres 1B |
Gene Aliases: |
2810458H16Rik |
Location: |
Chr.17:55651190-55712647 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pot1b | NM_028370.1 | NP_082646.1 | ||
| AK045286.1 | ||||
| AK050727.1 | ||||
| AK165040.1 | BAE38013.1 |
More Information
Set Membership: |
|

Genomic Map